ENA946841347 Enamine

1-(aminomethyl)-N,N-dimethylcyclopropane-1-sulfonamide hydrochloride

  • MDL number MFCD30734230
  • Cas Number : 2094319-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384380
Availability details for
Quantity
  • Availability
  • Add to Cart