ENA946841350 Enamine

1-azabicyclo[3.3.1]nonan-3-one hydrochloride

  • MDL number MFCD30734234
  • Cas Number : 90154-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384385
Availability details for
Quantity
  • Availability
  • Add to Cart