ENA946841356 Enamine

methyl 5-amino-4-chloro-2-acetamidobenzoate

  • MDL number MFCD30734241
  • Cas Number : 2095410-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384473
Availability details for
Quantity
  • Availability
  • Add to Cart