ENA946841367 Enamine

ethyl 6-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2-carboxylate hydrochloride

  • MDL number MFCD30734257
  • Cas Number : 2094209-62-6
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384620
Availability details for
Quantity
  • Availability
  • Add to Cart