ENA946841375 Enamine

2-[1-(bromomethyl)cyclobutyl]oxane

  • MDL number MFCD30734266
  • Cas Number : 2095409-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384671
Availability details for
Quantity
  • Availability
  • Add to Cart