ENA946841383 Enamine

acetic acid; tert-butyl 3-carbamimidoylpiperidine-1-carboxylate

  • MDL number MFCD30734277
  • Cas Number : 2138127-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CH3COOH
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384728
Availability details for
Quantity
  • Availability
  • Add to Cart