ENA946841387 Enamine

cyclohept-4-en-1-amine hydrochloride

  • MDL number MFCD30734281
  • Cas Number : 2094642-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-385210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385210
Availability details for
Quantity
  • Availability
  • Add to Cart