ENA946841388 Enamine

7-methyl-3-[(methylamino)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazin-8-one dihydrochloride

  • MDL number MFCD30734283
  • Cas Number : 2094628-99-4
  • Molecular Weight : 268.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-385232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385232
Availability details for
Quantity
  • Availability
  • Add to Cart