ENA946841389 Enamine

3-[(methylamino)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazin-8-one dihydrochloride

  • MDL number MFCD30734284
  • Cas Number : 2094616-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-385239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385239
Availability details for
Quantity
  • Availability
  • Add to Cart