ENA946841441 Enamine

2-(ethylamino)-N-methylacetamide hydrochloride

  • MDL number MFCD30734353
  • Cas Number : 149794-11-6
  • Molecular Weight : 152.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-386117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-386117
Availability details for
Quantity
  • Availability
  • Add to Cart