ENA946841447 Enamine

methyl({[6-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl})amine dihydrochloride

  • MDL number MFCD30734360
  • Cas Number : 2094926-49-3
  • Molecular Weight : 307.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-386195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-386195
Availability details for
Quantity
  • Availability
  • Add to Cart