ENA946841449 Enamine

(4-methoxy-5-methyl-1,3-thiazol-2-yl)methanol

  • MDL number MFCD30734362
  • Cas Number : 2095409-49-5
  • Molecular Weight : 159.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-386199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-386199
Availability details for
Quantity
  • Availability
  • Add to Cart