ENA946841894 Enamine

1-tert-butyl 3-chloromethyl azetidine-1,3-dicarboxylate

  • MDL number MFCD30734817
  • Cas Number : 2137597-32-9
  • Molecular Weight : 249.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-386779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-386779
Availability details for
Quantity
  • Availability
  • Add to Cart