ENA946842203 Enamine

[1-(4-bromophenyl)-1H-pyrazol-3-yl]methanamine

  • MDL number MFCD21304203
  • Cas Number : 1472723-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384354
Availability details for
Quantity
  • Availability
  • Add to Cart