ENA946842207 Enamine

4-(ethylamino)-N-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)benzene-1-sulfonamide

  • MDL number MFCD25142350
  • Cas Number : 1521098-42-9
  • Molecular Weight : 282.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-385278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385278
Availability details for
Quantity
  • Availability
  • Add to Cart