ENA946842211 Enamine

[2-(3,4-dichlorophenoxy)-5-fluorophenyl]methanol

  • MDL number MFCD12902168
  • Cas Number : 1179661-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-385293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385293
Availability details for
Quantity
  • Availability
  • Add to Cart