ENA946842214 Enamine

4-(cyclopropylmethyl)-6-(methylsulfanyl)-1,3,5-triazin-2-amine

  • MDL number MFCD25985388
  • Cas Number : 1557037-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384521
Availability details for
Quantity
  • Availability
  • Add to Cart