ENA946842242 Enamine

(2,1,3-benzothiadiazol-4-yl)methanamine hydrochloride

  • MDL number MFCD06411538
  • Cas Number : 830330-21-7
  • Molecular Weight : 201.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384377
Availability details for
Quantity
  • Availability
  • Add to Cart