ENA946842266 Enamine

2-[(1H-pyrazol-1-yl)methyl]pyridin-4-amine

  • MDL number MFCD21834435
  • Cas Number : 1541829-45-1
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384627
Availability details for
Quantity
  • Availability
  • Add to Cart