ENA946842270 Enamine

3-bromo-5-nitropyridine-4-carboxylic acid

  • MDL number MFCD28746363
  • Cas Number : 1805472-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384629
Availability details for
Quantity
  • Availability
  • Add to Cart