ENA946842293 Enamine

5-[(2,6-dichlorobenzenesulfonyl)methyl]furan-2-carboxylic acid

  • MDL number MFCD13719366
  • Cas Number : 897765-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-386130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-386130
Availability details for
Quantity
  • Availability
  • Add to Cart