ENA946842294 Enamine

6-[4-(propan-2-yl)phenoxy]pyridine-2,3-diamine

  • MDL number MFCD23350172
  • Cas Number : 1536771-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384333
Availability details for
Quantity
  • Availability
  • Add to Cart