ENA946842309 Enamine

4-(benzyloxy)-2,2-dimethylbutan-1-ol

  • MDL number MFCD30270446
  • Cas Number : 370555-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384462
Availability details for
Quantity
  • Availability
  • Add to Cart