ENA946842311 Enamine

7H-purine-6-carbohydrazide

  • MDL number MFCD20669479
  • Cas Number : 98198-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384463
Availability details for
Quantity
  • Availability
  • Add to Cart