ENA946842326 Enamine

7-methoxy-1,2,3,4-tetrahydroisoquinolin-6-ol

  • MDL number MFCD01444686
  • Cas Number : 1011-43-4
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384725

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-384725
pictograms GHS07
hstatements H302,H315,H319,H335,H412
Availability details for
Quantity
  • Availability
  • Add to Cart