ENA946842335 Enamine

1-bromo-3-(chloromethyl)naphthalene

  • MDL number MFCD18410255
  • Cas Number : 1261469-74-2
  • Molecular Weight : 255.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-384482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-384482
Availability details for
Quantity
  • Availability
  • Add to Cart