ENA946844724 Enamine

4-methylcycloheptan-1-one

  • MDL number MFCD24394757
  • Cas Number : 5452-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85702
Availability details for
Quantity
  • Availability
  • Add to Cart