ENA947386003 Enamine

N-[2-(1H-indol-3-yl)ethyl]benzenesulfonamide

  • MDL number MFCD04616178
  • Cas Number : 32585-54-9
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15902
Availability details for
Quantity
  • Availability
  • Add to Cart