ENA947386358 Enamine

5-[4-(propan-2-yl)phenyl]imidazolidine-2,4-dione

  • MDL number MFCD06364466
  • Cas Number : 1008672-28-3
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-17826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17826
Availability details for
Quantity
  • Availability
  • Add to Cart