ENA947386878 Enamine

adamantane

  • Cas Number : 281-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-212
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-19684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-19684
Availability details for
Quantity
  • Availability
  • Add to Cart