ENA947387177 Enamine

2,3-dimethylcyclohexan-1-amine

  • MDL number MFCD00001493
  • Cas Number : 42195-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 51
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-20570

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-20570
pictograms GHS02,GHS05
hstatements H226,H314
Availability details for
Quantity
  • Availability
  • Add to Cart