ENA947387414 Enamine

bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

  • Cas Number : 120-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-43
Purity 95
Flash Point 113
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21280

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-21280
s_phrases_eu 26-36/37/39-45
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart