ENA947387825 Enamine

N,N-diethyl-3,5-dimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD07366356
  • Cas Number : 944416-16-4
  • Molecular Weight : 231.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-35267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35267
Availability details for
Quantity
  • Availability
  • Add to Cart