ENA947388058 Enamine

N-(4-methylphenyl)-2-(1H-1,2,4-triazol-1-yl)acetamide

  • MDL number MFCD19445628
  • Cas Number : 1252441-64-7
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67645
Availability details for
Quantity
  • Availability
  • Add to Cart