ENA947388129 Enamine

3-amino-4,4,4-trifluorobutanenitrile

  • MDL number MFCD22524940
  • Cas Number : 1391333-68-8
  • Molecular Weight : 138.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-88330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88330
Availability details for
Quantity
  • Availability
  • Add to Cart