ENA947388931 Enamine

3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD31381137
  • Cas Number : 2126161-36-0
  • Molecular Weight : 312.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-412395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-412395
Availability details for
Quantity
  • Availability
  • Add to Cart