ENA947388961 Enamine

(2S)-2-[(4-bromophenoxy)methyl]pyrrolidine hydrochloride

  • MDL number MFCD30752557
  • Cas Number : 2126144-34-9
  • Molecular Weight : 292.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-624137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-624137
Availability details for
Quantity
  • Availability
  • Add to Cart