ENA947389356 Enamine

(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid

  • MDL number MFCD00155640
  • Cas Number : 144701-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-624040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-624040
Availability details for
Quantity
  • Availability
  • Add to Cart