ENA947389454 Enamine

2-(phenylamino)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD09878864
  • Cas Number : 133972-63-1
  • Molecular Weight : 220.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-624347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-624347
Availability details for
Quantity
  • Availability
  • Add to Cart