ENA947390429 Enamine

[5-(trifluoromethyl)oxolan-2-yl]methanol

  • MDL number MFCD31429593
  • Cas Number : 2138263-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-692423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-692423
Availability details for
Quantity
  • Availability
  • Add to Cart