ENA947391026 Enamine

2-(2-cyclopropyl-1,3-oxazol-5-yl)acetonitrile

  • MDL number MFCD28550622
  • Cas Number : 1935492-67-3
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-649225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-649225
Availability details for
Quantity
  • Availability
  • Add to Cart