ENA947391034 Enamine

1-[1-(3,5-dichlorophenyl)-1H-1,2,3-triazol-4-yl]-2,2,2-trifluoroethan-1-one

  • MDL number MFCD17362472
  • Cas Number : 1272305-66-4
  • Molecular Weight : 310.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-659025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-659025
Availability details for
Quantity
  • Availability
  • Add to Cart