ENA947391572 Enamine

tert-butyl 3-amino-2-(oxan-4-yl)propanoate

  • MDL number MFCD31420807
  • Cas Number : 2138394-93-9
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-251839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-251839
Availability details for
Quantity
  • Availability
  • Add to Cart