ENA947391627 Enamine

methyl 4-hydroxy-1-(pyrazin-2-yl)pyrrolidine-2-carboxylate

  • MDL number MFCD31381252
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-311656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-311656
Availability details for
Quantity
  • Availability
  • Add to Cart