ENA947392073 Enamine

6-chloro-3-{[2-(propan-2-yl)-1,3-thiazol-4-yl]methyl}-3,4-dihydroquinazolin-4-one

  • MDL number MFCD18120744
  • Cas Number : 1241067-67-3
  • Molecular Weight : 319.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-309506

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-309506
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart