ENA947392815 Enamine

4-methyl-4-(piperidin-3-yl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD30750061
  • Cas Number : 2126178-01-4
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-378132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-378132
Availability details for
Quantity
  • Availability
  • Add to Cart