ENA947392920 Enamine

sodium 2-[5-(oxan-4-yl)-4H-1,2,4-triazol-3-yl]acetate

  • MDL number MFCD30750696
  • Cas Number : 2094381-30-1
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-385132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-385132
Availability details for
Quantity
  • Availability
  • Add to Cart