ENA947392922 Enamine

2,5-diethyl 1,3,4-thiadiazole-2,5-dicarboxylate

  • MDL number MFCD31381283
  • Cas Number : 2126178-33-2
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-316213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-316213
Availability details for
Quantity
  • Availability
  • Add to Cart