ENA947392929 Enamine

tert-butyl 5,8-diazaspiro[3.6]decane-8-carboxylate

  • MDL number MFCD31381287
  • Cas Number : 2091455-57-9
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-368248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-368248
Availability details for
Quantity
  • Availability
  • Add to Cart