ENA947393031 Enamine

methyl 6-cyano-1,2,3,4-tetrahydronaphthalene-1-carboxylate

  • MDL number MFCD30750010
  • Cas Number : 2126161-98-4
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-367065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367065
Availability details for
Quantity
  • Availability
  • Add to Cart